missing translation for 'onlineSavingsMsg'
Learn More

1,10-Phenanthroline Monohydrochloride Monohydrate, 97%, Thermo Scientific Chemicals

Product Code. 10629263
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18851-33-7
Molecular Formula HCl·H2O
Molecular Weight (g/mol) 234.69
MDL Number MFCD00150061
InChI Key NDLHUHRGAIHALB-UHFFFAOYSA-N
Synonym 1,10-phenanthroline hydrochloride monohydrate, 1,10-phenanthroline hydrochloride hydrate, 1,10-phenanthroline monohydrochloride monohydrate, 1,10-phenanthroline hydrate hydrochloride, 1,10-phenanthroline, monohydrochloride, monohydrate, phen hydrate hydrochloride, acmc-209iz9, dsstox_cid_24309, dsstox_rid_80145, dsstox_gsid_44309
PubChem CID 2723715
IUPAC Name 1,10-phenanthroline;hydrate;hydrochloride
SMILES C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.O.Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 224.0°C to 225.0°C
Color Purple or White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 100 g
Solubility Information Solubility in water: freely soluble.
Formula Weight 234.69
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1, 10-Phenanthroline monohydrochloride monohydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,10-Phenanthroline monohydrochloride monohydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'