Learn More
1,2,2,6,6-Pentamethyl-4-piperidinol, 99%, Thermo Scientific Chemicals
19.10 € - 149.75 €
Chemical Identifiers
CAS | 2403-89-6 |
---|---|
Molecular Formula | C10H21NO |
Molecular Weight (g/mol) | 171.28 |
MDL Number | MFCD00191220 |
InChI Key | NWHNXXMYEICZAT-UHFFFAOYSA-N |
Synonym | 1,2,2,6,6-pentamethyl-4-piperidinol, 4-piperidinol, 1,2,2,6,6-pentamethyl, 4-hydroxy-1,2,2,6,6-pentamethylpiperidine, unii-lr520bam5v, 1,2,2,6,6-pentamethyl-4-hydroxypiperidine, lr520bam5v, methyl-taa-ol, hopemp, pubchem22251, acmc-209g8a |
PubChem CID | 75472 |
IUPAC Name | 1,2,2,6,6-pentamethylpiperidin-4-ol |
SMILES | CC1(CC(CC(N1C)(C)C)O)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10058521
|
Thermo Scientific Acros
300860050 |
5 g | Glass bottle |
19.10 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10421301
|
Thermo Scientific Acros
300862500 |
250 g | Plastic bottle |
47.03 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10043420
|
Thermo Scientific Acros
300860010 |
1 kg | Plastic bottle |
149.75 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2403-89-6 | |
171.28 | |
NWHNXXMYEICZAT-UHFFFAOYSA-N | |
75472 | |
CC1(CC(CC(N1C)(C)C)O)C |
C10H21NO | |
MFCD00191220 | |
1,2,2,6,6-pentamethyl-4-piperidinol, 4-piperidinol, 1,2,2,6,6-pentamethyl, 4-hydroxy-1,2,2,6,6-pentamethylpiperidine, unii-lr520bam5v, 1,2,2,6,6-pentamethyl-4-hydroxypiperidine, lr520bam5v, methyl-taa-ol, hopemp, pubchem22251, acmc-209g8a | |
1,2,2,6,6-pentamethylpiperidin-4-ol |
Specifications
2403-89-6 | |
100.0 | |
Yellow | |
238°C | |
Authentic | |
C10H21NO | |
0.967 | |
Solubility in water: in water: 4.8% (20°C). Other solubilities: in ethanol: 55% (20°C),in acetone: 38% (20°C),in chloroform: 52% (20°C) | |
CC1(CC(CC(N1C)(C)C)O)C | |
171.28 | |
15.3 mPa.s (80°C) | |
99% | |
1,2,2,6,6-Pentamethyl-4-piperidinol, 99% |
98.5 | |
70°C to 76°C | |
0.9670g/mL | |
118°C | |
99% | |
MFCD00191220 | |
1,2,2,6,6-pentamethyl-4-piperidinol, 4-piperidinol, 1,2,2,6,6-pentamethyl, 4-hydroxy-1,2,2,6,6-pentamethylpiperidine, unii-lr520bam5v, 1,2,2,6,6-pentamethyl-4-hydroxypiperidine, lr520bam5v, methyl-taa-ol, hopemp, pubchem22251, acmc-209g8a | |
NWHNXXMYEICZAT-UHFFFAOYSA-N | |
1,2,2,6,6-pentamethylpiperidin-4-ol | |
75472 | |
171.28 | |
Flakes |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 219-292-8
missing translation for 'tsca' : TSCA
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.