missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Tetrahydro-2,2,4,7-tetramethylquinoline, 97%, Thermo Scientific Chemicals

Product Code. 11392699
Click to view available options
Quantity:
5 mL
25 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59388-58-8
Molecular Formula C13H19N
Molecular Weight (g/mol) 189.302
MDL Number MFCD00799587
InChI Key NRWNXIXJZMSDAU-UHFFFAOYSA-N
PubChem CID 101035
IUPAC Name 2,2,4,7-tetramethyl-3,4-dihydro-1H-quinoline
SMILES CC1CC(NC2=C1C=CC(=C2)C)(C)C

Specifications

Density 0.975
Boiling Point 123°C (5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5475
Quantity 25 mL
Synonym 1,2,3,4-tetrahydro-2,2,4,7-tetramethylquinoline, 2,2,4,7-tetramethyl-1,2,3,4-tetrahydroquinoline, quinoline, 1,2,3,4-tetrahydro-2,2,4,7-tetramethyl, acmc-20ap7g, quinoline,1,2,3,4-tetrahydro-2,2,4,7-tetramethyl
Formula Weight 189.3
Percent Purity 97%
Chemical Name or Material 1,2,3,4-Tetrahydro-2,2,4,7-tetramethylquinoline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.