missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Tetrahydroquinoline, 98%, Thermo Scientific Chemicals

45.23 € - 148.74 €

Chemical Identifiers

CAS 635-46-1
Molecular Formula C9H11N
Molecular Weight (g/mol) 133.19
MDL Number MFCD00006693
InChI Key LBUJPTNKIBCYBY-UHFFFAOYSA-N
Synonym tetrahydroquinoline, 1,2,3,4-tetrahydro-quinoline, kusol, quinoline, 1,2,3,4-tetrahydro, unii-ccr50n1z9g, 1,2,3,4-tetrahydrochinoline, 1,2,3,4-tetrahydro quinoline, benzopiperidine, ccr50n1z9g, 1,2,3,4,-tetrahydroquinoline
PubChem CID 69460
ChEBI CHEBI:213323
IUPAC Name 1,2,3,4-tetrahydroquinoline
SMILES C1CNC2=CC=CC=C2C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11459767
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B22750.22
100 g
45.23 €
100g
Please to purchase this item. Need a web account? Register with us today!
11469767
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B22750.36
500 g
148.74 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

1,2,3,4-Tetrahydroquinoline is used as an antioxidant and corrosion inhibitor. It is an active component in various dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,3,4-Tetrahydroquinoline is used as an antioxidant and corrosion inhibitor. It is an active component in various dyes.

Solubility
Miscible with alcohol, ether, carbon disulfide and many organic solvents. Slightly miscible with water.

Notes
Hygroscopic. Incomaptible with strong oxidizing agents.
Specifications

Chemical Identifiers

635-46-1
133.19
LBUJPTNKIBCYBY-UHFFFAOYSA-N
69460
1,2,3,4-tetrahydroquinoline
C9H11N
MFCD00006693
tetrahydroquinoline, 1,2,3,4-tetrahydro-quinoline, kusol, quinoline, 1,2,3,4-tetrahydro, unii-ccr50n1z9g, 1,2,3,4-tetrahydrochinoline, 1,2,3,4-tetrahydro quinoline, benzopiperidine, ccr50n1z9g, 1,2,3,4,-tetrahydroquinoline
CHEBI:213323
C1CNC2=CC=CC=C2C1

Specifications

635-46-1
1.059
100°C (212°F)
1.595
116149
Miscible with alcohol,ether,carbon disulfide and many organic solvents. Slightly miscible with water.
C1CNC2=CC=CC=C2C1
133.19
CHEBI:213323
99%
∼14°C
248°C to 250°C
C9H11N
MFCD00006693
tetrahydroquinoline, 1,2,3,4-tetrahydro-quinoline, kusol, quinoline, 1,2,3,4-tetrahydro, unii-ccr50n1z9g, 1,2,3,4-tetrahydrochinoline, 1,2,3,4-tetrahydro quinoline, benzopiperidine, ccr50n1z9g, 1,2,3,4,-tetrahydroquinoline
LBUJPTNKIBCYBY-UHFFFAOYSA-N
1,2,3,4-tetrahydroquinoline
69460
133.19
1,2,3,4-Tetrahydroquinoline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c

H315-H319-H335-H350

missing translation for 'einecsNumber' : 211-237-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only