missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Tetrahydroquinoxaline, 98%, Thermo Scientific Chemicals

Product Code. 15441738
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3476-89-9
Molecular Formula C8H10N2
Molecular Weight (g/mol) 134.18
MDL Number MFCD00047564
InChI Key HORKYAIEVBUXGM-UHFFFAOYSA-N
Synonym 1,2,3,4-tetrahydro-quinoxaline, tetrahydroquinoxaline, quinoxaline, 1,2,3,4-tetrahydro, 1,3,4-tetrahydroquinoxaline, ksc495i0b, 1,2,3,4-terahydroquinoxaline, quinoxaline,2,3,4-tetrahydro, 1,2,3,4-tetrahydro quinoxaline, benzo 2,3 piperazine, tetrahydro
PubChem CID 77028
IUPAC Name 1,2,3,4-tetrahydroquinoxaline
SMILES C1CNC2=CC=CC=C2N1

Specifications

Melting Point 92°C to 98°C
Quantity 5 g
Formula Weight 134.18
Percent Purity 98%
Chemical Name or Material 1,2,3,4-Tetrahydroquinoxaline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'