missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,6-Tetrahydropyridine, 98%, Thermo Scientific Chemicals

110.55 € - 371.85 €

Chemical Identifiers

CAS 694-05-3
Molecular Formula C5H9N
Molecular Weight (g/mol) 83.13
MDL Number MFCD00005949
InChI Key FTAHXMZRJCZXDL-UHFFFAOYSA-N
Synonym pyridine, 1,2,3,6-tetrahydro, 3-piperideine, 1,2,5,6-tetrahydropyridine, unii-26rls9d255, delta 3-piperideine, .delta.3-piperidine, delta3-piperidine, acmc-1bcyc, 1,3,6-tetrahydropyridine, 1,5,6-tetrahydropyridine
PubChem CID 12750
ChEBI CHEBI:47860
IUPAC Name 1,2,3,6-tetrahydropyridine
SMILES C1CNCC=C1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10585134
View Documents
Thermo Scientific Acros
158870010
1 g Glass bottle
110.55 €
1g
Please to purchase this item. Need a web account? Register with us today!
10654935
View Documents
Thermo Scientific Acros
158870050
5 g Glass bottle
371.85 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

694-05-3
83.13
FTAHXMZRJCZXDL-UHFFFAOYSA-N
12750
1,2,3,6-tetrahydropyridine
C5H9N
MFCD00005949
pyridine, 1,2,3,6-tetrahydro, 3-piperideine, 1,2,5,6-tetrahydropyridine, unii-26rls9d255, delta 3-piperideine, .delta.3-piperidine, delta3-piperidine, acmc-1bcyc, 1,3,6-tetrahydropyridine, 1,5,6-tetrahydropyridine
CHEBI:47860
C1CNCC=C1

Specifications

694-05-3
100.0
Cream-Yellow to White
108.0°C
Authentic
C5H9N
MFCD00005949
0.91
Solubility in water: soluble.
C1CNCC=C1
83.13
CHEBI:47860
98%
1,2,3,6-Tetrahydropyridine
97.5
-48.0°C
0.9100g/mL
16°C
97.5% min. (GC)
1.48
20, III, 1912
pyridine, 1,2,3,6-tetrahydro, 3-piperideine, 1,2,5,6-tetrahydropyridine, unii-26rls9d255, delta 3-piperideine, .delta.3-piperidine, delta3-piperidine, acmc-1bcyc, 1,3,6-tetrahydropyridine, 1,5,6-tetrahydropyridine
FTAHXMZRJCZXDL-UHFFFAOYSA-N
1,2,3,6-tetrahydropyridine
12750
83.13
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If eye irritation persists: Get medical advice/a

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 211-766-2

Product Identifier
  • 1,2,3,6-Tetrahydropyridine

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.