missing translation for 'onlineSavingsMsg'
Learn More

1,2,3-Heptanetriol, 95%, high melting isomer, mixture of diastereoisomers, Thermo Scientific Chemicals

Product Code. 10005443 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 103404-57-5
CAS Min % 94.0
CAS Max % 100.0
Melting Point 92.0°C to 97.0°C
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Molecular Formula C7H16O3
Linear Formula CH3(CH2)3CH(OH)CH(OH)CH2OH
MDL Number MFCD00010517
Quantity 1 g
Synonym 1,2,3-heptanetriol, 1,2,3-trihydroxyheptane, 1-butylglycerol, 1-n-butylglycerol, 1,2,3-heptanetriol isomer t 1 m solution, 1,2,3-heptanetriol isomer h 0.5 solution, acmc-1c7bt, 1,2,3-heptanetriol solution, additive screening solution 08/fluka kit no 78374, additive screening solution 09/fluka kit no 78374
InChI Key HXYCHJFUBNTKQR-UHFFFAOYSA-N
SMILES CCCCC(C(CO)O)O
IUPAC Name heptane-1,2,3-triol
Molecular Weight (g/mol) 148.2
PubChem CID 124604
ChEBI CHEBI:67095
Formula Weight 148.2
Percent Purity 95%
Chemical Name or Material 1, 2, 3-Heptanetriol
Show More Show Less
compliance-icons
Product Identifier
  • 1,2,3-Heptanetriol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.