missing translation for 'onlineSavingsMsg'
Learn More

1,2,3-Thiadiazole-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10374392
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 4100-13-4
Molecular Formula C3H2N2O2S
Molecular Weight (g/mol) 130.121
MDL Number MFCD00052099
InChI Key HJZYBDPHAHGHAZ-UHFFFAOYSA-N
Synonym 1,2,3-thiadiazole-4-carboxylic acid, 1,2,3 thiadiazole-4-carboxylic acid, thiadiazolecarboxylicacid, 4-carboxy-1,2,3-thiadiazole, 1,2,3-thiadiazole4-carboxylic acid, 1 pound not2 pound not3 thiadiazole-4-carboxylicacid
PubChem CID 351418
IUPAC Name thiadiazole-4-carboxylic acid
SMILES C1=C(N=NS1)C(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 130.13
Percent Purity 97%
Chemical Name or Material 1, 2, 3-Thiadiazole-4-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.