missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Triazin-3-amine, 97%, Thermo Scientific™

Product Code. 10479644
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1120-99-6
Molecular Formula C3H4N4
Molecular Weight (g/mol) 96.093
InChI Key MJIWQHRXSLOUJN-UHFFFAOYSA-N
Synonym 3-amino-1,2,4-triazine, as-triazine, 3-amino, 3-amino-as-triazine, 1,2,4-triazin-3-ylamine, 1,2,4-triazine-3-ylamine, 1,2,4 triazin-3-ylamine, zlchem 1277, 1,2,4-triazine-3-amine, 3-amino-1,2,4,-triazine
PubChem CID 70715
IUPAC Name 1,2,4-triazin-3-amine
SMILES C1=CN=NC(=N1)N

Specifications

Assay Percent Range 97%
Quantity 5 g
Formula Weight 96.09
Chemical Name or Material 1, 2, 4-Triazin-3-amine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'