missing translation for 'onlineSavingsMsg'
Learn More
1,2,4-Triazole, 99%, Thermo Scientific Chemicals Product Code.: 11494673

1,2,4-Triazole, 99%, Thermo Scientific Chemicals

Product Code. 11494673
500 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11494673

Brand: Thermo Scientific Alfa Aesar A11597.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1,2,4-Triazole is widely used as an intermediate in the manufacture of pharmaceuticals. It acts as an anti-scaling agent and antifungal compound. It is widely used in antifungals such as fluconazole anditraconazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,4-Triazole is widely used as an intermediate in the manufacture of pharmaceuticals. It acts as an anti-scaling agent and antifungal compound. It is widely used in antifungals such as fluconazole anditraconazole.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 288-88-0
Molecular Formula C2H3N3
Molecular Weight (g/mol) 69.067
MDL Number MFCD00005228
InChI Key NSPMIYGKQJPBQR-UHFFFAOYSA-N
Synonym 1,2,4-triazole, 4h-1,2,4-triazole, pyrrodiazole, s-triazole, 1,2,4-1h-triazole, 1h-1,2,4-triazol, unii-10ms0y1rdi, 1,2,4 triazole, 1,2,4-triazol, 4h-1,2,4-triazole van
PubChem CID 9257
ChEBI CHEBI:35550
IUPAC Name 1H-1,2,4-triazole
SMILES C1=NC=NN1

Specifications

Melting Point 118°C to 122°C
Boiling Point 260°C
Flash Point 158°C (316°F)
Quantity 500 g
Beilstein 104767
Merck Index 14,9605
Solubility Information Soluble in water.
Formula Weight 69.07
Percent Purity 99%
Chemical Name or Material 1,2,4-Triazole

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ13E046 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA11597.22,A11597.14,A11597.36
Lot NumberY20A003 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA11597.22,A11597.14,A11597.36
Lot Number5001V01V Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA11597.22,A11597.14,A11597.36
Lot NumberI9712A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA11597.22,A11597.14,A11597.36
Lot NumberF19R037 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA11597.22,A11597.14,A11597.36
compliance-icons compliance-icons
Product Identifier
  • 1,2,4-Triazole
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : 1,2,4-Triazole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,2,4-Triazole, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.