missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,4-Trichlorobenzene, 99%, pure, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 16.0°C |
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Density | 1.4500g/mL |
Boiling Point | 214.0°C |
Quantity | 1 L |
Infrared Spectrum | Authentic |
Packaging | Glass Bottle |
Physical Form | Liquid |
Refractive Index | 1.5700 to 1.5725 |
Show More |
Safety and Handling
Product Identifier
- 1,2,4-Trichlorobenzene
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Skin corrosion/irritation Category 2
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H315-Causes skin irritation.
- H302-Harmful if swallowed.
- P273-Avoid release to the environment.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'