missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,4-Trichlorobenzene, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 g
2500 g
4 x 2.5 kg
Description
1,2,4-Trichlorobenzene is used as a dielectric and heat transfer fluid in transformers. It acts as an intermediate, degreaser, wood preservative and solvent for dye. It is a high-temperature solvent used in gel permeation chromatography, especially for polyethylene and polypropylene. Further, it is used as a lubricant and as a synthetic transformer oil.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,2,4-Trichlorobenzene is used as a dielectric and heat transfer fluid in transformers. It acts as an intermediate, degreaser, wood preservative and solvent for dye. It is a high-temperature solvent used in gel permeation chromatography, especially for polyethylene and polypropylene. Further, it is used as a lubricant and as a synthetic transformer oil.
Solubility
Miscible with ether, benzene, CS{2}.Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
1,2,4-Trichlorobenzene is used as a dielectric and heat transfer fluid in transformers. It acts as an intermediate, degreaser, wood preservative and solvent for dye. It is a high-temperature solvent used in gel permeation chromatography, especially for polyethylene and polypropylene. Further, it is used as a lubricant and as a synthetic transformer oil.
Solubility
Miscible with ether, benzene, CS{2}.Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
Melting Point | 17°C |
Density | 1.454g/mL |
Boiling Point | 213°C |
Quantity | 4 x 2.5 kg |
Flash Point | 110°C (230°F) |
Physical Form | Liquid |
Refractive Index | 1.572 |
UN Number | UN2321 |
Merck Index | 14,9631 |
Solubility Information | Miscible with ether,benzene,CS2;Immiscible with water. |
Show More |
Safety and Handling
Product Identifier
- 1,2,4-Trichlorobenzene
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Skin corrosion/irritation Category 2
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- MIXTURE LIST-Contains : 1,2,4-Trichlorobenzene
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'