missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Trichlorobenzene, ReagentPlus™, ≥99%, Honeywell™

Product Code. 15681270
Click to view available options
Quantity:
1 L
2.5 L
20 L
Packaging:
Glass Bottle
Stainless Steel Can
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-82-1
Molecular Formula C6H3Cl3
Molecular Weight (g/mol) 181.44
MDL Number MFCD00000547
InChI Key PBKONEOXTCPAFI-UHFFFAOYSA-N
Synonym benzene, 1,2,4-trichloro, unsym-trichlorobenzene, hostetex l-pec, trojchlorobenzen, 1,2,4-trichlorbenzol, 1,2,4-trichlorobenzol, 1,3,4-trichlorobenzene, trichlorobenzene a, 1,2,5-trichlorobenzene, as-trichlorobenzene
PubChem CID 13
ChEBI CHEBI:28222
IUPAC Name 1,2,4-trichlorobenzene
SMILES C1=CC(=C(C=C1Cl)Cl)Cl

Specifications

Melting Point 17°C
Color Colorless
Density 1.454g/mL at 25°C
Boiling Point 213.5°C
Quantity 1 L
Explosion Limit 6.6% (150°F)
Flash Point 110°C (230°F)
Ignition Point 1060°F
Packaging Glass Bottle
Physical Form Liquid
Refractive Index n20/D 1.571(lit.)
Vapor Pressure 1mmHg (40°C)
UN Number UN2321
Beilstein 956819
Formula Weight 181.45g/mol
Percent Purity ≥99%
Grade ReagentPlus™
Chemical Name or Material 1,2,4-Trichlorobenzene
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'