missing translation for 'onlineSavingsMsg'
Learn More
1,2:5,6-Di-O-isopropylidene-alpha-D-allofuranose, 98%, Thermo Scientific Chemicals Product Code.: 15457729

1,2:5,6-Di-O-isopropylidene-alpha-D-allofuranose, 98%, Thermo Scientific Chemicals

Product Code. 15457729
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15457729

Brand: Thermo Scientific Alfa Aesar J66388.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2595-05-3
MDL Number MFCD00135634
ChEBI CHEBI:34917

Specifications

Melting Point 73°C to 76°C
Color White
Molecular Formula C12H17O4PS2
Quantity 1 g
Beilstein 1345413
Synonym phenthoate, fenthoate, dimephenthioate, elsan, dimephenthoate, cidial, phendal, dimefenthoat, cidemul, papthion
InChI Key XAMUDJHXFNRLCY-UHFFFAOYNA-N
SMILES CCOC(=O)C(C1=CC=CC=C1)SP(=S)(OC)OC
IUPAC Name ethyl 2-dimethoxyphosphinothioylsulfanyl-2-phenylacetate
Molecular Weight (g/mol) 320.36
PubChem CID 17435
Formula Weight 260.28
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1,2:5,6-Di-O-isopropylidene-alpha-D-allofuranose
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,2:5,6-Di-O-isopropylidene-alpha-D-allofuranose, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.