missing translation for 'onlineSavingsMsg'
Learn More

1,2,5-Oxadiazole-3-carboxylic acid, ≥97%, Thermo Scientific™

Product Code. 10259292
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 88598-08-7
Molecular Formula C3H2N2O3
Molecular Weight (g/mol) 114.06
MDL Number MFCD00604386
InChI Key JBLHCUQCDKBPGY-UHFFFAOYSA-N
Synonym furazan-3-carboxylic acid, 3-carboxy-1,2,5-oxadiazole, 1,2,5-oxadiazole-3-carboxylicacid, acmc-1bkac, 1,2,5-oxadiazole, oxd4, 9ci
PubChem CID 1415524
IUPAC Name 1,2,5-oxadiazole-3-carboxylic acid
SMILES OC(=O)C1=NON=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 114.06
Percent Purity 97%
Chemical Name or Material 1, 2, 5-Oxadiazole-3-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'