missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(2-Aminoethyl)imidazolidin-2-one , Tech., Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Specifications
Specifications
Packaging | Amber glass bottle |
Quantity | 1 g |
Formula Weight | 129.16 |
Percent Purity | Tech. |
Grade | Technical |
Chemical Name or Material | 1-(2-Aminoethyl)imidazolidin-2-one |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
1-(2-Aminoethyl)imidazolidin-2-one , Tech., Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction