Learn More
1,2-Benzanthracene, 99%, Thermo Scientific Chemicals
41.91 € - 110.55 €
Chemical Identifiers
CAS | 56-55-3 |
---|---|
Molecular Formula | C18H12 |
Molecular Weight (g/mol) | 228.29 |
MDL Number | MFCD00003599 |
InChI Key | DXBHBZVCASKNBY-UHFFFAOYSA-N |
Synonym | benz a anthracene, tetraphene, 1,2-benzanthracene, benzanthrene, benzo a anthracene, benzanthracene, naphthanthracene, 2,3-benzphenanthrene, benzoanthracene |
PubChem CID | 5954 |
ChEBI | CHEBI:51348 |
IUPAC Name | benzo[a]anthracene |
SMILES | C1=CC=C2C=C3C(C=CC4=CC=CC=C34)=CC2=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10628124
|
Thermo Scientific Acros
105252500 |
250 mg | Glass bottle |
41.91 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10569800
|
Thermo Scientific Acros
105250010 |
1 g | Glass bottle |
110.55 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
56-55-3 | |
228.29 | |
DXBHBZVCASKNBY-UHFFFAOYSA-N | |
5954 | |
benzo[a]anthracene |
C18H12 | |
MFCD00003599 | |
benz a anthracene, tetraphene, 1,2-benzanthracene, benzanthrene, benzo a anthracene, benzanthracene, naphthanthracene, 2,3-benzphenanthrene, benzoanthracene | |
CHEBI:51348 | |
C1=CC=C2C=C3C(C=CC4=CC=CC=C34)=CC2=C1 |
Specifications
56-55-3 | |
100.0 | |
Beige to Green-Yellow | |
Authentic | |
C18H12 | |
05,718 | |
benz a anthracene, tetraphene, 1,2-benzanthracene, benzanthrene, benzo a anthracene, benzanthracene, naphthanthracene, 2,3-benzphenanthrene, benzoanthracene | |
DXBHBZVCASKNBY-UHFFFAOYSA-N | |
benzo[a]anthracene | |
5954 | |
228.29 | |
Crystalline Powder |
98.5 | |
158°C to 161°C | |
437.6°C | |
98.5% min. (GC) | |
MFCD00003599 | |
15,1064 | |
Solubility in water: insoluble in water. Other solubilities: difficulty soluble in boiling alcohol,soluble in most other common organic solvents | |
C1=CC=C2C=C3C(C=CC4=CC=CC=C34)=CC2=C1 | |
228.29 | |
CHEBI:51348 | |
99% | |
1, 2-Benzanthracene, 99% |
Safety and Handling
GHS H Statement:
Very toxic to aquatic life with long lasting effects.
May cause cancer.
GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 200-280-6
missing translation for 'rtecsNumber' : CV9275000
missing translation for 'tsca' : TSCA
- 1,2-Benzanthracene
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1B
Hazard Statement
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H350-May cause cancer.
Precautionary Statement
- P201-Obtain special instructions before use.
- P273-Avoid release to the environment.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only