missing translation for 'onlineSavingsMsg'
Learn More
1,2-Benzenedithiol, 96%, Thermo Scientific Chemicals Product Code.: 10125720

1,2-Benzenedithiol, 96%, Thermo Scientific Chemicals

Product Code. 10125720 Shop All Thermo Scientific Products
1 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10125720

Brand: Thermo Scientific 334920010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17534-15-5
Molecular Formula C6H6S2
Molecular Weight (g/mol) 142.23
MDL Number MFCD00004835
InChI Key JRNVQLOKVMWBFR-UHFFFAOYSA-N
Synonym 1,2-benzenedithiol, benzenedithiol, 1,2-dimercaptobenzene, dithiocatechol, dimercaptobenzene, o-benzenedithiol, acmc-209e9y, 1,2-benzenedithiol 9ci, benzene-1,2-dithiolShow MoreShow Less
PubChem CID 69370
IUPAC Name benzene-1,2-dithiol
SMILES SC1=C(S)C=CC=C1

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 27.0°C to 28.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.6565 to 1.6585
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 142.2
Percent Purity 96%
Physical Form Liquid After Melting
Chemical Name or Material 1, 2-Benzenedithiol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465338 Certificate TypeCertificate of Analysis Date21/08/2024 Catalog Number334920010,334920050
Lot NumberA0392083 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number334920010,334920050
Lot NumberA0406744 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number334920010,334920050
Lot NumberA0445243 Certificate TypeCertificate of Analysis Date29/08/2022 Catalog Number334920010,334920050
Lot NumberA0442592 Certificate TypeCertificate of Analysis Date03/06/2022 Catalog Number334920010,334920050
compliance-icons
Product Identifier
  • 1,2-Benzenedithiol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,2-Benzenedithiol, 96%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.