missing translation for 'onlineSavingsMsg'
Learn More
1,2-Bis((2S,5S)-2,5-diethylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate, 97%, Thermo Scientific Chemicals Product Code.: 10666840

1,2-Bis((2S,5S)-2,5-diethylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate, 97%, Thermo Scientific Chemicals

Product Code. 10666840
250 mg, Glass bottle
Click to view available options
Quantity:
100 mg
250 mg
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10666840

Brand: Thermo Scientific Acros 435192500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 213343-64-7
Molecular Formula C30H48BF4P2Rh
Molecular Weight (g/mol) 660.37
MDL Number MFCD01862465
InChI Key DSYBEQKPSKLNMC-RTXTXWNLNA-N
Synonym s,s-et-duphos-rh, +-1,2-bis 2s,5s-2,5-diethylphospholano benzene cyclooctadiene rhodium i tetrafluoroborate, +-1,2-bis 2s,5s-2,5-diethylphospholano benzene cyclooctadiene rhodium l tetrafluoroborateShow MoreShow Less
PubChem CID 11006841
SMILES [Rh+].F[B-](F)(F)F.C1C\C=C/CC\C=C/1.CC[C@H]1CC[C@H](CC)P1C1=C(C=CC=C1)P1[C@@H](CC)CC[C@@H]1CC

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Brown to Orange
Elemental Analysis 52.4 to 56.8% C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 250 mg
IUPAC Name (1Z,5Z)-cycloocta-1,5-diene;(2S,5S)-1-[2-[(2S,5S)-2,5-diethylphospholan-1-yl]phenyl]-2,5-diethylphospholane;rhodium;tetrafluoroborate
Formula Weight 660.37
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1,2-Bis((2S,5S)-2,5-Diethylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number342342 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number435192500,435191000
Product Identifier
  • 1,2-Bis((2S,5S)-2,5-diethylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,2-Bis((2S,5S)-2,5-diethylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.