missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(dicyclohexylphosphino)ethane, 98%, Thermo Scientific Chemicals

Product Code. 10146653
Click to view available options
Quantity:
1 g
5 g

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23743-26-2
Molecular Formula C26H48P2
Molecular Weight (g/mol) 422.61
MDL Number MFCD00015521
InChI Key BOUYBUIVMHNXQB-UHFFFAOYSA-N
Synonym 1,2-bis dicyclohexylphosphino ethane, ethylenebis dicyclohexylphosphine, phosphine, 1,2-ethanediylbis dicyclohexyl, 1,2-ethanediylbis dicyclohexyl phosphine, dicyclohexyl 2-dicyclohexylphosphanylethyl phosphane, bis 1,2-dicyclohexylphosphino ethane, pubchem6558, acmc-1capw, ethane-1,2-diylbis dicyclohexylphosphane, phosphine, 1,2-ethanediylbis*dicyclohexyl
PubChem CID 534202
IUPAC Name dicyclohexyl(2-dicyclohexylphosphanylethyl)phosphane
SMILES C1CCC(CC1)P(CCP(C2CCCCC2)C3CCCCC3)C4CCCCC4

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble in water
Formula Weight 422.61
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1, 2-Bis(dicyclohexylphosphino)ethane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,2-Bis(dicyclohexylphosphino)ethane
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H413-May cause long lasting harmful effects to aquatic life.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 1,2-Bis(dicyclohexylphosphino)ethane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.