missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Bis(dimethylphosphino)ethane, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
0.25 g
1 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 179°C to 180°C |
Density | 0.9 |
Boiling Point | 180°C |
Flash Point | −17°C (1.4°F) |
Odor | Recognizable |
Linear Formula | (CH3)2PCH2CH2P(CH3)2 |
Refractive Index | 1.507 |
Quantity | 1 g |
UN Number | UN1993 |
Sensitivity | Air Sensitive |
Show More |
Safety and Handling
Product Identifier
- 1,2-Bis(dimethylphosphino)ethane
- Danger
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : 1,2-Bis(dimethylphosphino)ethane
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'