missing translation for 'onlineSavingsMsg'
Learn More

1-[2-(Boc-amino)ethyl]indole-6-carboxylic acid, 97%, Thermo Scientific™

Product Code. 15428416
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 885266-67-1
Molecular Formula C9H5N3O2S
Molecular Weight (g/mol) 219.218
MDL Number MFCD04973974
InChI Key FDQWHYSXVRDVMV-UHFFFAOYSA-N
Synonym 4-nitro-1h-indol-3-yl sulfanyl formonitrile
PubChem CID 24720942
IUPAC Name (4-nitro-1H-indol-3-yl) thiocyanate
SMILES C1=CC2=C(C(=C1)[N+](=O)[O-])C(=CN2)SC#N

Specifications

Color White
Quantity 250 mg
Formula Weight 304.34
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-[2-(Boc-amino)ethyl]indole-6-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'