missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Bromoethoxy)-4-chlorobenzene, 99%, Thermo Scientific Chemicals

162.00 € - 460.00 €

Chemical Identifiers

CAS 2033-76-3
Molecular Formula C8H8BrClO
Molecular Weight (g/mol) 235.51
MDL Number MFCD00000617
InChI Key YYFLBDSMQRWARK-UHFFFAOYSA-N
Synonym 1-2-bromoethoxy-4-chlorobenzene, benzene, 1-2-bromoethoxy-4-chloro, 2-bromoethyl 4-chlorophenyl ether, 4-chlorophenyl 2-bromoethyl ether, 2-4-chlorophenoxy ethyl bromide, benzene,1-2-bromoethoxy-4-chloro, 2-bromo-1-4-chlorophenoxy ethane, pubchem17076, acmc-1cjap, p-chlorophenoxyethyl bromide
PubChem CID 74854
IUPAC Name 1-(2-bromoethoxy)-4-chlorobenzene
SMILES C1=CC(=CC=C1OCCBr)Cl
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10421743
View Documents
Thermo Scientific Acros
379830050
5 g Glass bottle
162.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
10056622
View Documents
Thermo Scientific Acros
379830250
25 g Glass bottle
460.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2033-76-3
235.51
YYFLBDSMQRWARK-UHFFFAOYSA-N
74854
C1=CC(=CC=C1OCCBr)Cl
C8H8BrClO
MFCD00000617
1-2-bromoethoxy-4-chlorobenzene, benzene, 1-2-bromoethoxy-4-chloro, 2-bromoethyl 4-chlorophenyl ether, 4-chlorophenyl 2-bromoethyl ether, 2-4-chlorophenoxy ethyl bromide, benzene,1-2-bromoethoxy-4-chloro, 2-bromo-1-4-chlorophenoxy ethane, pubchem17076, acmc-1cjap, p-chlorophenoxyethyl bromide
1-(2-bromoethoxy)-4-chlorobenzene

Specifications

2033-76-3
100.0
150°C to 152°C (18.0 mmHg)
Authentic
C8H8BrClO
1.5570 to 1.5590 (20°C, 589nm)
1-2-bromoethoxy-4-chlorobenzene, benzene, 1-2-bromoethoxy-4-chloro, 2-bromoethyl 4-chlorophenyl ether, 4-chlorophenyl 2-bromoethyl ether, 2-4-chlorophenoxy ethyl bromide, benzene,1-2-bromoethoxy-4-chloro, 2-bromo-1-4-chlorophenoxy ethane, pubchem17076, acmc-1cjap, p-chlorophenoxyethyl bromide
C1=CC(=CC=C1OCCBr)Cl
235.51
235.51
Liquid or Low Melting Solid
98.5
38°C to 39°C
150°C
98.5% min. (GC)
ClC6H4O(CH2)2Br
MFCD00000617
YYFLBDSMQRWARK-UHFFFAOYSA-N
1-(2-bromoethoxy)-4-chlorobenzene
74854
99%
1-(2-Bromoethoxy)-4-chlorobenzene
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if inhaled.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 217-994-9

Product Identifier
  • 1-(2-Bromoethoxy)-4-chlorobenzene

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.