missing translation for 'onlineSavingsMsg'
Learn More

1,2-Cyclohexanediol, 98%, mixture of cis and trans, Thermo Scientific Chemicals

Product Code. 10743641
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 931-17-9
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00003861
InChI Key PFURGBBHAOXLIO-UHFFFAOYSA-N
Synonym 1,2-cyclohexanediol, 2-hydroxycyclohexanol, pyrocatechitol, 1,2-dihydroxycyclohexane, brenzkatechin, 1,2-benzenediol, hexahydro, brenzkatechin german, 1,2-trans-cyclohexanediol, trans-1,2-dihydroxycyclohexane, 1,2-cyclohexanediol, mixture of cis and trans
PubChem CID 13601
ChEBI CHEBI:24567
IUPAC Name cyclohexane-1,2-diol
SMILES C1CCC(C(C1)O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 73°C to 77°C
Color White
Boiling Point 118°C to 120°C (10.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic bottle
Linear Formula C6H10(OH)2
Quantity 25 g
Beilstein 06, 740
Solubility Information Solubility in water: soluble
Formula Weight 116.16
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 1, 2-Cyclohexanediol, 98%
Show More Show Less
Product Identifier
  • 1,2-Cyclohexanediol, mixture of cis and trans

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'