missing translation for 'onlineSavingsMsg'
Learn More

1,2-Cyclohexanedione, 98%, Thermo Scientific Chemicals

Product Code. 10385980
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 765-87-7
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.13
InChI Key OILAIQUEIWYQPH-UHFFFAOYSA-N
Synonym 1,2-cyclohexanedione, 1,2-dioxocyclohexane, cyclohexanedione, 1,2-cyclohexadione, cyclohexan-1,2-dione, ccris 6296, unii-75c1ovw0fj, 75c1ovw0fj, 1,2-cyclohexanedione,ketone form, dihydrocatechol
PubChem CID 13006
ChEBI CHEBI:41674
IUPAC Name cyclohexane-1,2-dione
SMILES C1CCC(=O)C(=O)C1

Specifications

Melting Point 35.0°C to 38.0°C
Color Yellow
Boiling Point 193.0°C to 195.0°C
Flash Point 84°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H8(=O)2
Refractive Index 1.5180 to 1.5200
Quantity 50 g
Beilstein 07,II,526
Fieser 02,93
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol and diethyl ether
Formula Weight 112.13
Percent Purity 98%
Physical Form Liquid After Melting
Chemical Name or Material 1, 2-Cyclohexanedione
Show More Show Less
compliance-icons
Product Identifier
  • 1,2-Cyclohexanedione
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.