missing translation for 'onlineSavingsMsg'
Learn More

1,2-Cyclohexanedione dioxime, 97%, Thermo Scientific Chemicals

Product Code. 11429240
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1,2-Cyclohexanedione dioxime acts as a chelating ligand forming complexes with many transition metal ions like Co, Fe and Ni. It is used in photometric detection of metal ions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Cyclohexanedione dioxime acts as a chelating ligand forming complexes with many transition metal ions like Co, Fe and Ni. It is used in photometric detection of metal ions.

Solubility
Soluble in water.

Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 492-99-9
Molecular Formula C6H10N2O2
Molecular Weight (g/mol) 142.16
MDL Number MFCD00001663
InChI Key CUNNCKOPAWXYDX-KQQUZDAGSA-N
Synonym 1,2-cyclohexanedione dioxime, nioxime, 1z,2z-cyclohexane-1,2-dione dioxime, nioxim, 1,2-cyclohexanedione, dioxime, 1,2-bis hydroxyimino cyclohexane, unii-g1qt7su5a8, cyclohexane-1,2-dione dioxime, 1,2-cyclohexanedione, 1,2-dioxime, g1qt7su5a8
PubChem CID 10300
IUPAC Name N-(2-nitrosocyclohexen-1-yl)hydroxylamine
SMILES O\N=C1/CCCC/C/1=N\O

Specifications

Melting Point 181°C to 188°C (decomposition)
Quantity 100 g
Beilstein 1909047
Merck Index 14,6560
Solubility Information Soluble in water.
Formula Weight 142.16
Percent Purity 97%
Chemical Name or Material 1,2-Cyclohexanedione dioxime

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.