missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Diaminocyclohexanetetraacetic acid monohydrate, Honeywell Fluka™
Click to view available options
Quantity:
25 g
100 g
6 x 25 g
6 x 100 g
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
Specifications
Quantity | 25 g |
UN Number | NONH for all modes of transport |
Beilstein | 6884761 |
Formula Weight | 364.35g/mol |
Percent Purity | ≥98.5% |
Chemical Name or Material | 1,2-Diaminocyclohexanetetraacetic acid monohydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'