missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Diaminoethane, Anhydrous, SLR, Fisher Chemical
Click to view available options
2.5L, Plastic Coated Glass bottle
Specifications
Specifications
CAS | 107-15-3 |
Melting Point | 11°C |
Color | Colorless |
pH | 12.2 |
Boiling Point | 117°C |
Packaging | Plastic Coated Glass bottle |
Molecular Formula | C2H8N2 |
MDL Number | 8204 |
Quantity | 2.5 L |
Formula Weight | 60.1g/mol |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction