missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Diaminopropane, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
250 mL
500 mL
1000 mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | ∼-37°C |
Density | 0.869 |
Boiling Point | 119°C to 120°C |
Flash Point | 34°C (93°F) |
Odor | Characteristic |
Refractive Index | 1.446 |
Quantity | 500 mL |
UN Number | UN2258 |
Beilstein | 605274 |
Sensitivity | Air sensitive; Hygroscopic |
Show More |
Safety and Handling
Product Identifier
- 1,2-Diaminopropane
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 A
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : 1,2-Diaminopropane
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'