missing translation for 'onlineSavingsMsg'
Learn More

1,2-Diaminopropane, 99%, Thermo Scientific Chemicals

Product Code. 10458233
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-90-0
Molecular Formula C3H10N2
Molecular Weight (g/mol) 74.13
MDL Number MFCD00008089
InChI Key AOHJOMMDDJHIJH-UHFFFAOYNA-N
Synonym 1,2-diaminopropane, propylenediamine, 1,2-propanediamine, propylene diamine, 2,3-diaminopropane, 1,2-propylenediamine, 1,2 diaminopropane, ccris 4863, 1-methylethylenediamine, 2-aminopropylamine
PubChem CID 6567
ChEBI CHEBI:30630
IUPAC Name propane-1,2-diamine
SMILES CC(N)CN

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -37.0°C
Density 0.8700g/mL
Boiling Point 119.0°C to 120.0°C
Flash Point 33°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH(NH2)CH2NH2
Refractive Index 1.4450 to 1.4470
Quantity 250 mL
Beilstein 04,257
Merck Index 15,7965
Specific Gravity 0.87
Solubility Information Solubility in water: soluble. Other solubilities: insoluble in ether, soluble in acetone, benzene, chloroform and, ethanol
Formula Weight 74.13
Percent Purity 99%
Chemical Name or Material 1, 2-Diaminopropane
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1,2-Diaminopropane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H311-Toxic in contact with skin.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'