Learn More
1,2-Dianilinoethane, 97%, Thermo Scientific Chemicals
24.90 € - 223.00 €
Chemical Identifiers
CAS | 150-61-8 |
---|---|
Molecular Formula | C14H16N2 |
Molecular Weight (g/mol) | 212.30 |
MDL Number | MFCD00003019 |
InChI Key | NOUUUQMKVOUUNR-UHFFFAOYSA-N |
Synonym | 1,2-dianilinoethane, n,n'-diphenylethylenediamine, stabilite, n,n'-ethylenedianiline, nodx, n1,n2-diphenylethane-1,2-diamine, sym-diphenylethylenediamine, 1,2-ethanediamine, n,n'-diphenyl, aniline, n,n'-ethylenedi, n,n'-difenylethylendiamin |
PubChem CID | 67422 |
IUPAC Name | N,N'-diphenylethane-1,2-diamine |
SMILES | C(CNC1=CC=CC=C1)NC1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11466296
|
Thermo Scientific Alfa Aesar
B23098.06 |
5 g |
24.90 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11476296
|
Thermo Scientific Alfa Aesar
B23098.14 |
25 g |
78.50 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11486296
|
Thermo Scientific Alfa Aesar
B23098.22 |
100 g |
223.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1,2-Dianilinoethane can forms crystalline imidazolidines with aldehydes in the presence of AcOH. N-donating ligand. Can be used as a trapping agent for aldehydes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,2-Dianilinoethane can forms crystalline imidazolidines with aldehydes in the presence of AcOH. N-donating ligand. Can be used as a trapping agent for aldehydes.
Solubility
Sparingly soluble in water 0.072 g/L @ 25°C.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Chemical Identifiers
150-61-8 | |
212.30 | |
NOUUUQMKVOUUNR-UHFFFAOYSA-N | |
67422 | |
C(CNC1=CC=CC=C1)NC1=CC=CC=C1 |
C14H16N2 | |
MFCD00003019 | |
1,2-dianilinoethane, n,n'-diphenylethylenediamine, stabilite, n,n'-ethylenedianiline, nodx, n1,n2-diphenylethane-1,2-diamine, sym-diphenylethylenediamine, 1,2-ethanediamine, n,n'-diphenyl, aniline, n,n'-ethylenedi, n,n'-difenylethylendiamin | |
N,N'-diphenylethane-1,2-diamine |
Specifications
150-61-8 | |
228°C (12 mmHg) | |
MFCD00003019 | |
14,2990 | |
Sparingly soluble in water 0.072g/L @ 25°C. | |
C(CNC1=CC=CC=C1)NC1=CC=CC=C1 | |
212.30 | |
212.3 | |
1,2-Dianilinoethane |
64°C to 68°C | |
C14H16N2 | |
646740 | |
1,2-dianilinoethane, n,n'-diphenylethylenediamine, stabilite, n,n'-ethylenedianiline, nodx, n1,n2-diphenylethane-1,2-diamine, sym-diphenylethylenediamine, 1,2-ethanediamine, n,n'-diphenyl, aniline, n,n'-ethylenedi, n,n'-difenylethylendiamin | |
NOUUUQMKVOUUNR-UHFFFAOYSA-N | |
N,N'-diphenylethane-1,2-diamine | |
67422 | |
≥98% |
Safety and Handling
GHS H Statement
H301-H311-H332
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'einecsNumber' : 205-765-6
missing translation for 'rtecsNumber' : KV4800000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.