missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromo-1,2-diphenylethane, 96%, Thermo Scientific Chemicals

38.50 € - 107.00 €

Chemical Identifiers

CAS 5789-30-0
Molecular Formula C14H12Br2
Molecular Weight (g/mol) 340.05
MDL Number MFCD00000137
InChI Key GKESIQQTGWVOLH-UHFFFAOYSA-N
Synonym 1,2-dibromo-1,2-diphenylethane, stilbene dibromide, 1,2-dibromo-2-phenylethyl benzene, benzene, 1,1'-1,2-dibromo-1,2-ethanediyl bis, meso-dibromostilbene, benzene, 1,1'-1,2-dibromo-1,2-ethanediyl bis-, r*,s*, alpha,alpha-dibromobibenzyl, bibenzyl,, a,, a'-dibromo, bibenzyl, alpha,alpha'-dibromo, acmc-1ao3z
PubChem CID 93010
IUPAC Name (1,2-dibromo-2-phenylethyl)benzene
SMILES C1=CC=C(C=C1)C(C(C2=CC=CC=C2)Br)Br
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10245050
View Documents
Thermo Scientific Acros
147950050
5 g Glass bottle
38.50 €
5g
Please to purchase this item. Need a web account? Register with us today!
10055733
View Documents
Thermo Scientific Acros
147950250
25 g Glass bottle
107.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5789-30-0
340.05
GKESIQQTGWVOLH-UHFFFAOYSA-N
93010
C1=CC=C(C=C1)C(C(C2=CC=CC=C2)Br)Br
C14H12Br2
MFCD00000137
1,2-dibromo-1,2-diphenylethane, stilbene dibromide, 1,2-dibromo-2-phenylethyl benzene, benzene, 1,1'-1,2-dibromo-1,2-ethanediyl bis, meso-dibromostilbene, benzene, 1,1'-1,2-dibromo-1,2-ethanediyl bis-, r*,s*, alpha,alpha-dibromobibenzyl, bibenzyl,, a,, a'-dibromo, bibenzyl, alpha,alpha'-dibromo, acmc-1ao3z
(1,2-dibromo-2-phenylethyl)benzene

Specifications

5789-30-0
100.0
Authentic
C14H12Br2
MFCD00000137
1,2-dibromo-1,2-diphenylethane, stilbene dibromide, 1,2-dibromo-2-phenylethyl benzene, benzene, 1,1'-1,2-dibromo-1,2-ethanediyl bis, meso-dibromostilbene, benzene, 1,1'-1,2-dibromo-1,2-ethanediyl bis-, r*,s*, alpha,alpha-dibromobibenzyl, bibenzyl,, a,, a'-dibromo, bibenzyl, alpha,alpha'-dibromo, acmc-1ao3z
C1=CC=C(C=C1)C(C(C2=CC=CC=C2)Br)Br
340.05
340.05
Powder
95.0
235°C to 241°C
95% min. (HPLC)
C6H5CH(Br)CH(Br)C6H5
05, 602
GKESIQQTGWVOLH-UHFFFAOYSA-N
(1,2-dibromo-2-phenylethyl)benzene
93010
96%
1, 2-Dibromo-1, 2-diphenylethane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 227-324-7

Product Identifier
  • 1,2-Dibromo-1,2-diphenylethane

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only