Learn More
1,2-Dibromo-2-methylpropane, 98%, Thermo Scientific Chemicals
24.92 € - 490.44 €
Chemical Identifiers
CAS | 594-34-3 |
---|---|
Molecular Formula | C4H8Br2 |
Molecular Weight (g/mol) | 215.92 |
MDL Number | MFCD00000126 |
InChI Key | SDTXSEXYPROZSZ-UHFFFAOYSA-N |
Synonym | isobutylene bromide, 1,2-dibromoisobutane, acmc-1aolc, propane,2-dibromo-2-methyl, propane,1,2-dibromo-2-methyl, 1,2-dibromo-2-methylpropane |
PubChem CID | 222657 |
IUPAC Name | 1,2-dibromo-2-methylpropane |
SMILES | CC(C)(CBr)Br |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10156790
|
Thermo Scientific Acros
406490050 |
5 g | Glass bottle |
24.92 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10647181
|
Thermo Scientific Acros
406490250 |
25 g | Glass bottle |
71.56 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10240303
|
Thermo Scientific Acros
406492500 |
250 g | Glass bottle |
490.44 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
594-34-3 | |
215.92 | |
SDTXSEXYPROZSZ-UHFFFAOYSA-N | |
222657 | |
CC(C)(CBr)Br |
C4H8Br2 | |
MFCD00000126 | |
isobutylene bromide, 1,2-dibromoisobutane, acmc-1aolc, propane,2-dibromo-2-methyl, propane,1,2-dibromo-2-methyl, 1,2-dibromo-2-methylpropane | |
1,2-dibromo-2-methylpropane |
Specifications
594-34-3 | |
100.0 | |
1.7900g/mL | |
>110°C | |
97.5% min. (GC) | |
(CH3)2CBrCH2Br | |
MFCD00000126 | |
isobutylene bromide, 1,2-dibromoisobutane, acmc-1aolc, propane,2-dibromo-2-methyl, propane,1,2-dibromo-2-methyl, 1,2-dibromo-2-methylpropane | |
SDTXSEXYPROZSZ-UHFFFAOYSA-N | |
1,2-dibromo-2-methylpropane | |
222657 | |
98% | |
1, 2-Dibromo-2-methylpropane, 98% |
97.5 | |
Colorless | |
150.0°C | |
Authentic | |
C4H8Br2 | |
1.5083 to 1.5103 | |
1.79 | |
Solubility in water: slightly soluble | |
CC(C)(CBr)Br | |
215.92 | |
215.92 | |
Liquid |
Safety and Handling
GHS H Statement:
Suspected of causing cancer.
GHS P Statement:
Use personal protective equipment as required.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 209-835-7
missing translation for 'rtecsNumber' : TX8810000
missing translation for 'tsca' : TSCA
- 1,2-Dibromo-2-methylpropane
Signal Word
- Warning
Hazard Category
- Carcinogenicity Category 2
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H351-Suspected of causing cancer.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.