missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromobutane, 98%, Thermo Scientific Chemicals

48.24 € - 501.50 €

Chemical Identifiers

CAS 533-98-2
Molecular Formula C4H8Br2
Molecular Weight (g/mol) 215.92
MDL Number MFCD00000157
InChI Key CZWSZZHGSNZRMW-UHFFFAOYNA-N
Synonym butane, 1,2-dibromo, .alpha.-butylene dibromide, alpha-butylene dibromide, 1,2-dibomobutane, 1,2 dibromobutane, butane,2-dibromo, 1,2-dibromo-butane, alpha-butylene bromide, 1,2-butylene bromide, .alpha.-butylene bromide
PubChem CID 10792
IUPAC Name 1,2-dibromobutane
SMILES CCC(Br)CBr
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11494247
View Documents
Thermo Scientific Alfa Aesar
B20356.18
50 g
48.24 €
50g
Please to purchase this item. Need a web account? Register with us today!
11404257
View Documents
Thermo Scientific Alfa Aesar
B20356.30
250 g
165.00 €
250g
Please to purchase this item. Need a web account? Register with us today!
11484247
View Documents
Thermo Scientific Alfa Aesar
B20356.0B
1000 g
501.50 €
1000g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

533-98-2
215.92
CZWSZZHGSNZRMW-UHFFFAOYNA-N
10792
CCC(Br)CBr
C4H8Br2
MFCD00000157
butane, 1,2-dibromo, .alpha.-butylene dibromide, alpha-butylene dibromide, 1,2-dibomobutane, 1,2 dibromobutane, butane,2-dibromo, 1,2-dibromo-butane, alpha-butylene bromide, 1,2-butylene bromide, .alpha.-butylene bromide
1,2-dibromobutane

Specifications

533-98-2
1.789
>110°C (230°F)
1.5141
1731380
butane, 1,2-dibromo, .alpha.-butylene dibromide, alpha-butylene dibromide, 1,2-dibomobutane, 1,2 dibromobutane, butane,2-dibromo, 1,2-dibromo-butane, alpha-butylene bromide, 1,2-butylene bromide, .alpha.-butylene bromide
CCC(Br)CBr
215.92
215.93
1,2-Dibromobutane
-65°C
166°C to 167°C
C4H8Br2
MFCD00000157
14,1566
CZWSZZHGSNZRMW-UHFFFAOYNA-N
1,2-dibromobutane
10792
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III

missing translation for 'einecsNumber' : 208-581-4

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only