missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Dichlorobenzene, 99+%, for HPLC, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
1 L
Packaging:
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | -15.0°C |
Density | 1.3000g/mL |
Boiling Point | 179.0°C to 180.0°C |
Flash Point | 67°C |
Assay Percent Range | 99% min. (GC) |
Packaging | Glass Bottle |
Linear Formula | C6H4Cl2 |
Refractive Index | 1.5500 to 1.5520 |
Quantity | 1 L |
Merck Index | 15,3065 |
Show More |
Safety and Handling


Product Identifier
- 1,2-Dichlorobenzene
- Warning
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H410-Very toxic to aquatic life with long lasting effects.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
1,2-Dichlorobenzene, 99+%, for HPLC, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction