missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichlorobenzene, 99%, pure, Thermo Scientific Chemicals

Product Code. 10751734
Click to view available options
Quantity:
5 mL
1 L
2.5 L
25 L
Packaging:
Glass Bottle
Metal drum

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-50-1
MDL Number MFCD00000535
InChI Key RFFLAFLAYFXFSW-UHFFFAOYSA-N
Synonym o-dichlorobenzene, chloroben, o-dichlorbenzol, dilantin db, ortho-dichlorobenzene, 2-dichlorobenzene, o-dichlorobenzol, odcb, benzene, 1,2-dichloro, cloroben
PubChem CID 7239
ChEBI CHEBI:35290
IUPAC Name 1,2-dichlorobenzene
SMILES C1=CC=C(C(=C1)Cl)Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -15.0°C
Color Colorless to Yellow
Density 1.3000g/mL
Boiling Point 179.0°C to 180.0°C
Flash Point 67°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Metal drum
Molecular Formula C6H4Cl2
Linear Formula C6H4Cl2
Refractive Index 1.5500 to 1.5520
Quantity 25 L
Beilstein 05, 201
Merck Index 15, 3065
Specific Gravity 1.3
Solubility Information Solubility in water: 0.13g/L(20°C). Other solubilities: miscible with most common organic solvents,miscible with alcohol,ether and benzene
Molecular Weight (g/mol) 147
Formula Weight 147
Vapor Pressure 1.3mbar at 20°C
Percent Purity 99%
Grade Pure
Physical Form Liquid
Chemical Name or Material 1, 2-Dichlorobenzene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'