missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichlorobenzene, 99%, Thermo Scientific Chemicals

Product Code. 11411578
Click to view available options
Quantity:
10,000 g
2500 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1,2-Dichlorobenzene is widely used as a precursor to prepare 1,2-dichloro-4-nitrobenzene, which is an intermediate in the synthesis of agrochemicals. It acts as a solvent for fullerenes. It is also used in softening and removing carbon-based contamination on metal surfaces. It also used as food preservative, motor oil additive, lubricant and paint.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Dichlorobenzene is widely used as a precursor to prepare 1,2-dichloro-4-nitrobenzene, which is an intermediate in the synthesis of agrochemicals. It acts as a solvent for fullerenes. It is also used in softening and removing carbon-based contamination on metal surfaces. It also used as food preservative, motor oil additive, lubricant and paint.

Solubility
Miscible with water and most of the organic solvents.

Notes
Keep away from sources of ignition. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-50-1
Molecular Formula C6H4Cl2
Molecular Weight (g/mol) 146.998
MDL Number MFCD00000535
InChI Key RFFLAFLAYFXFSW-UHFFFAOYSA-N
Synonym o-dichlorobenzene, chloroben, o-dichlorbenzol, dilantin db, ortho-dichlorobenzene, 2-dichlorobenzene, o-dichlorobenzol, odcb, benzene, 1,2-dichloro, cloroben
PubChem CID 7239
ChEBI CHEBI:35290
IUPAC Name 1,2-dichlorobenzene
SMILES C1=CC=C(C(=C1)Cl)Cl

Specifications

Melting Point -18°C
Density 1.306
Boiling Point 180°C
Flash Point 68°C (154°F)
Odor Pungent
Refractive Index 1.551
Quantity 10,000 g
UN Number UN1591
Beilstein 606078
Merck Index 14,3056
Solubility Information Miscible with water and most of the organic solvents.
Formula Weight 147
Percent Purity 99%
Chemical Name or Material 1,2-Dichlorobenzene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'