Learn More
1,2-Dichlorobenzene, Spectrophotometric Grade, 98%, Thermo Scientific Chemicals
Maximum level of impurities: Evaporation residue 5ppm, H2O 0.02%
73.87 € - 285.42 €
Chemical Identifiers
CAS | 95-50-1 |
---|---|
Molecular Formula | C6H4Cl2 |
Molecular Weight (g/mol) | 146.998 |
MDL Number | MFCD00000535 |
InChI Key | RFFLAFLAYFXFSW-UHFFFAOYSA-N |
Synonym | o-dichlorobenzene, chloroben, o-dichlorbenzol, dilantin db, ortho-dichlorobenzene, 2-dichlorobenzene, o-dichlorobenzol, odcb, benzene, 1,2-dichloro, cloroben |
PubChem CID | 7239 |
ChEBI | CHEBI:35290 |
IUPAC Name | 1,2-dichlorobenzene |
SMILES | C1=CC=C(C(=C1)Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11348337
|
Thermo Scientific Alfa Aesar
032154.K2 |
1 L |
73.87 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11328337
|
Thermo Scientific Alfa Aesar
032154.M1 |
4 x 1 L |
285.42 €
4 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11338337
|
Thermo Scientific Alfa Aesar
032154.K7 |
4 L |
239.19 €
4 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1,2-Dichlorobenzene is used in spectrophotometry and environmental testing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
95-50-1 | |
146.998 | |
RFFLAFLAYFXFSW-UHFFFAOYSA-N | |
7239 | |
1,2-dichlorobenzene |
C6H4Cl2 | |
MFCD00000535 | |
o-dichlorobenzene, chloroben, o-dichlorbenzol, dilantin db, ortho-dichlorobenzene, 2-dichlorobenzene, o-dichlorobenzol, odcb, benzene, 1,2-dichloro, cloroben | |
CHEBI:35290 | |
C1=CC=C(C(=C1)Cl)Cl |
Specifications
95-50-1 | |
Colorless | |
180°C | |
640°C (1184°F) | |
Pungent | |
1.551 | |
606078 | |
Miscible with water and most of the organic solvents. | |
C1=CC=C(C(=C1)Cl)Cl | |
146.998 | |
CHEBI:35290 | |
1.33 hPa (1mm Hg) at 20°C | |
1, 2-Dichlorobenzene | |
1, 2-Dichlorobenzene, Filtered through 0.2μ filters. |
°C to 18°C | |
1.306 | |
68°C (154°F) | |
98% | |
C6H4Cl2 | |
MFCD00000535 | |
o-dichlorobenzene, chloroben, o-dichlorbenzol, dilantin db, ortho-dichlorobenzene, 2-dichlorobenzene, o-dichlorobenzol, odcb, benzene, 1,2-dichloro, cloroben | |
RFFLAFLAYFXFSW-UHFFFAOYSA-N | |
1,2-dichlorobenzene | |
7239 | |
147 | |
Spectrophotometric | |
Liquid |
Safety and Handling
This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
P210-P235-P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P370+P378q-P501c
H227-H302+H332-H315-H317-H319-H335
missing translation for 'dotInformation' : DOT Class: 6.1, Packaging Group: III
missing translation for 'einecsNumber' : 202-425-9
missing translation for 'rtecsNumber' : CZ4500000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only