missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichloroethane, ≥99.0%, ACS Reagent, Honeywell™ Riedel-de Haën™

Product Code. 15663600
Click to view available options
Quantity:
500 mL
Packaging:
Glass Bottle
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

ACS Reagent,≥99.0%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Specifications

Melting Point -35°C
CAS 107-06-2
Density 1.256g/mL at 25°C
Boiling Point 83°C
Quantity 500 mL
Explosion Limit 0.156
Flash Point 13°C (55.4°F)
Ignition Point 775°F
Packaging Glass Bottle
Molecular Formula C2H4Cl2
Linear Formula ClCH2CH2Cl
MDL Number MFCD00000963
Vapor Pressure 87mmHg (25°C)
UN Number UN1184
Beilstein 605264
Synonym ethylene dichloride, ethylene chloride, ethane, 1,2-dichloro, dutch liquid, dutch oil, ethane dichloride, glycol dichloride, aethylenchlorid, dichloro-1,2-ethane, dichloremulsion
InChI Key WSLDOOZREJYCGB-UHFFFAOYSA-N
SMILES ClCCCl
IUPAC Name 1,2-dichloroethane
Molecular Weight (g/mol) 98.95
PubChem CID 11
ChEBI CHEBI:27789
Formula Weight 98.96g/mol
Percent Purity ≥99.0%
Grade ACS Reagent
Chemical Name or Material 1,2-Dichloroethane
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'