Learn More
1,2-Dichloroethane, 99.8%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
77.08 € - 316.58 €
Chemical Identifiers
CAS | 107-06-2 |
---|---|
Molecular Formula | C2H4Cl2 |
Molecular Weight (g/mol) | 98.95 |
MDL Number | MFCD00000963 |
InChI Key | WSLDOOZREJYCGB-UHFFFAOYSA-N |
Synonym | ethylene dichloride, ethylene chloride, ethane, 1,2-dichloro, dutch liquid, dutch oil, ethane dichloride, glycol dichloride, aethylenchlorid, dichloro-1,2-ethane, dichloremulsion |
PubChem CID | 11 |
ChEBI | CHEBI:27789 |
IUPAC Name | 1,2-dichloroethane |
SMILES | ClCCCl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10333532
|
Thermo Scientific Acros
326841000 |
100 mL | AcroSeal™ Glass Bottle |
77.08 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10612033
|
Thermo Scientific Acros
326840010 |
1 L | AcroSeal™ Glass Bottle |
155.78 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11924471
|
Thermo Scientific Acros
326840025 |
2.5 L | AcroSeal™ Glass Bottle |
316.58 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
107-06-2 | |
98.95 | |
WSLDOOZREJYCGB-UHFFFAOYSA-N | |
11 | |
1,2-dichloroethane |
C2H4Cl2 | |
MFCD00000963 | |
ethylene dichloride, ethylene chloride, ethane, 1,2-dichloro, dutch liquid, dutch oil, ethane dichloride, glycol dichloride, aethylenchlorid, dichloro-1,2-ethane, dichloremulsion | |
CHEBI:27789 | |
ClCCCl |
Specifications
-35.0°C | |
99.75 | |
100.0 | |
81.0°C to 85.0°C | |
Authentic | |
1.4440 to 1.4460 | |
0.0005 % max. | |
C2H4Cl2 | |
MFCD00000963 | |
0.005% Max. (Coulometric) | |
Solubility in water: 8.7g/L (20°C). Other solubilities: miscible with most common organic solvents | |
ClCCCl | |
98.95 | |
CHEBI:27789 | |
99.8% | |
1,2-Dichloroethane |
107-06-2 | |
Colorless | |
1.2500g/mL | |
13°C | |
Liquid | |
99.8% | |
1.25 | |
ClCH2CH2Cl | |
0.8 mPa.s (20°C) | |
ethylene dichloride, ethylene chloride, ethane, 1,2-dichloro, dutch liquid, dutch oil, ethane dichloride, glycol dichloride, aethylenchlorid, dichloro-1,2-ethane, dichloremulsion | |
WSLDOOZREJYCGB-UHFFFAOYSA-N | |
1,2-dichloroethane | |
11 | |
98.96 | |
Water <50ppm |
Safety and Handling
GHS H Statement:
May cause cancer.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
Harmful if swallowed.
GHS P Statement:
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
missing translation for 'einecsNumber' : 203-458-1
missing translation for 'rtecsNumber' : KI0525000
missing translation for 'tsca' : TSCA
- 1,2-Dichloroethane
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Acute toxicity Category 3
- Specific target organ toxicity Category 3
- Carcinogenicity Category 1B
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H350-May cause cancer.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P311-Call a POISON CENTER/doctor/ .
- P362-Take off contaminated clothing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.