missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichloroethane, Extra Pure, SLR, Fisher Chemical™

31.75 € - 430.00 €

Chemical Identifiers

CAS 107-06-2
Molecular Formula C2H4Cl2
Molecular Weight (g/mol) 98.95
MDL Number 963
InChI Key WSLDOOZREJYCGB-UHFFFAOYSA-N
IUPAC Name 1,2-dichloroethane
SMILES ClCCCl
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10754751
SDS View Documents
Fisher Chemical
D/1750/08
500 mL Amber Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10374930
SDS View Documents
Fisher Chemical
D/1750/17
2.5 L Amber Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10734561
SDS View Documents
Fisher Chemical
D/1750/21
5 L Metal Can This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10569950
SDS View Documents
Fisher Chemical
D/1750/25
25 L Metal Can
430.00 €
25 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

107-06-2
98.95
WSLDOOZREJYCGB-UHFFFAOYSA-N
ClCCCl
C2H4Cl2
963
1,2-dichloroethane

Specifications

-35°C
Colorless
Liquid
963
0.8 mPaS at 20°C
ClCCCl
98.95
107-06-2
81°C
C2H4Cl2
65mmHg 29
WSLDOOZREJYCGB-UHFFFAOYSA-N
1,2-dichloroethane
98.96g/mol
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 1,2-Dichloroethane

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Carcinogenicity Category 1B

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H350-May cause cancer.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ .

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.