missing translation for 'onlineSavingsMsg'
Learn More

1,2-Diiodoethane, 99%, Thermo Scientific Chemicals

Product Code. 10580561
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 624-73-7
Molecular Formula C2H4I2
Molecular Weight (g/mol) 281.85
MDL Number MFCD00001093
InChI Key GBBZLMLLFVFKJM-UHFFFAOYSA-N
Synonym ethane, 1,2-diiodo, ethylene diiodide, ethylene iodide, 1,2-diiodethane, 1,2-diiodo ethane, 1,2-bis iodanyl ethane, ch2ich2i, 1,2-diiodoethane, acmc-1b2u1, ksc358c5b
PubChem CID 12224
IUPAC Name 1,2-diiodoethane
SMILES C(CI)I

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 80.0°C to 83.0°C
Color Brown
Density 2.1300g/mL
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula ICH2CH2I
Quantity 25 g
Beilstein 01, 99
Specific Gravity 2.13
Formula Weight 281.85
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 1, 2-Diiodoethane, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 1,2-Diiodoethane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'