missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Dimethoxyethane, ≥99%, Reagent Grade, Inhibitor-free, Honeywell™
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
20 L
Packaging:
Glass bottle
Stainless steel can
Description
- ≥95% for general laboratory use.

Specifications
Specifications
Melting Point | -58°C |
Color | Colorless |
Density | 0.867g/mL at 25°C |
pH | Neutral |
Boiling Point | 85°C (1013 hPa) |
Flash Point | −2°C (28.4°F) |
Packaging | Stainless steel can |
Linear Formula | CH3OCH2CH2OCH3 |
Quantity | 20 L |
UN Number | UN2252 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
1,2-Dimethoxyethane, ≥99%, Reagent Grade, Inhibitor-free, Honeywell™ >
Spot an opportunity for improvement?Share a Content Correction