missing translation for 'onlineSavingsMsg'
Learn More

1,2-Ethanedisulfonic acid dihydrate, 98%, Thermo Scientific Chemicals

Product Code. 10486333
25 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5982-56-9
Molecular Formula C2H6O6S2·2H2O
Molecular Weight (g/mol) 226.22
MDL Number MFCD00069901
InChI Key DSGUSEBCDAKBCM-UHFFFAOYSA-N
Synonym 1,2-ethanedisulfonic acid dihydrate, unii-0j653yp17s, 1,2-ethanedisulfonic acid, dihydrate, 1,2-ethanedisulfonic acid dihydrate mi, 1,2-ethanedisulfonic acid, hydrate 1:2, ethane disulfonic acid dihydrate, ethane-1,2-disulfonic acid dihydrate, ethane-1,2-disulfonic acid-water 1/2
PubChem CID 57346009
IUPAC Name ethane-1,2-disulfonic acid;dihydrate
SMILES C(CS(=O)(=O)O)S(=O)(=O)O.O.O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 109°C to 113°C
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Beilstein 04,II,528
Merck Index 15, 3779
Solubility Information Solubility in water: very soluble
Formula Weight 226.22
Percent Purity 98%
Chemical Name or Material 1, 2-Ethanedisulfonic acid dihydrate
Show More Show Less
compliance-icons
Product Identifier
  • 1,2-Ethanedisulfonic acid dihydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,2-Ethanedisulfonic acid dihydrate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.