missing translation for 'onlineSavingsMsg'
Learn More

1-[2-Methoxy-5-(trifluoromethoxy)phenyl]ethanol, 95%, Thermo Scientific™

Product Code. 15497885
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 468074-91-1
Molecular Formula C10H11F3O3
Molecular Weight (g/mol) 236.19
MDL Number MFCD08062374
InChI Key DKUPMKWSJITPFC-UHFFFAOYNA-N
Synonym 1-2-methoxy-5-trifluoromethoxy phenyl ethan-1-ol, 1-2-methoxy-5-trifluoromethoxy phenyl ethanol, 1-hydroxy-1-2-methoxy-5-triflouromethoxy phenyl ethane
PubChem CID 18542515
IUPAC Name 1-[2-methoxy-5-(trifluoromethoxy)phenyl]ethanol
SMILES COC1=C(C=C(OC(F)(F)F)C=C1)C(C)O

Specifications

Quantity 1 g
Formula Weight 236.19
Percent Purity 95%
Chemical Name or Material 1-[2-Methoxy-5-(trifluoromethoxy)phenyl]ethanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'