missing translation for 'onlineSavingsMsg'
Learn More

1,2-Pentanediol, 96%, Thermo Scientific Chemicals

Product Code. 15264614
Click to view available options
Quantity:
50 mL
250 mL
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5343-92-0
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.15
MDL Number MFCD00010736
InChI Key WCVRQHFDJLLWFE-UHFFFAOYNA-N
Synonym 1,2-pentanediol, 1,2-dihydroxypentane, 1,2-pentanediol, 2r, acmc-20mbh5, acmc-1axdb, 3-01-00-02191 beilstein handbook reference, ksc271i3n, wcvrqhfdjllwfe-uhfffaoysa
PubChem CID 93000
IUPAC Name pentane-1,2-diol
SMILES CCCC(O)CO

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9710g/mL
Boiling Point 206.0°C
Flash Point 105°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)2CH(OH)CH2OH
Refractive Index 1.4387 to 1.4407
Quantity 50 mL
Beilstein 01,II,548
Specific Gravity 0.971
Solubility Information Solubility in water: miscible.
Formula Weight 104.15
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 1, 2-Pentanediol, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 1,2-Pentanediol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P403-Store in a well-ventilated place.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Pentane-1,2-diol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.