Learn More
1,2-Propanediol, ACS, 99.5% min
Beschreibung
1,2-Propanediol, ACS is used as a substitute for ethylene glycol and glycerol. It is a good solvent for resins. It dissolves many essential oils but is immiscible with fixed oils.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Spezifikation
| CAS | 57-55-6 |
| Melting Point | °C to 60°C |
| Density | 1.036 |
| Boiling Point | 186°C to 188°C |
| Flash Point | 107°C (225°F) |
| Assay Percent Range | 99.5% |
| Molecular Formula | C3H8O2 |
| Refractive Index | 1.432 |
| MDL Number | MFCD00064272 |
| Quantity | 500 mL |
| Mehr anzeigen |
RUO – Research Use Only
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.