missing translation for 'onlineSavingsMsg'
Learn More

1-[3-(4-chlorophenyl)-5-methylisoxazol-4-yl]ethan-1-one, 97%, Thermo Scientific™

20.90 € - 320.00 €

Chemical Identifiers

CAS 169814-48-6
Molecular Formula C12H10ClNO2
Molecular Weight (g/mol) 235.67
MDL Number MFCD01313507
InChI Key XSAMMAPYCPHHKH-UHFFFAOYSA-N
Synonym 1-3-4-chlorophenyl-5-methylisoxazol-4-yl ethan-1-one, 1-3-4-chlorophenyl-5-methylisoxazol-4-yl ethanone, 1-3-4-chlorophenyl-5-methyl-1,2-oxazol-4-yl ethanone, 3-4-chlorophenyl-4-acetyl-5-methylisoxazole, ethanone, 1-3-4-chlorophenyl-5-methyl-4-isoxazolyl, maybridge1_008689, 4-acetyl-3-4-chlorophenyl-5-methylisoxazole, 1-3-4-chlorophenyl-5-methyl-4-isoxazolyl-ethanone, 1-3-4-chloro-phenyl-5-methyl-isoxazol-4-yl-ethanone
PubChem CID 2735240
IUPAC Name 1-[3-(4-chlorophenyl)-5-methyl-1,2-oxazol-4-yl]ethan-1-one
SMILES CC(=O)C1=C(C)ON=C1C1=CC=C(Cl)C=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10652083
View Documents
Thermo Scientific Maybridge
SPB04957DA
1 g
20.90 €
1g
Please to purchase this item. Need a web account? Register with us today!
10385202
View Documents
Thermo Scientific Maybridge
SPB04957EA
10 g
150.00 €
10g
Please to purchase this item. Need a web account? Register with us today!
10311785
View Documents
Thermo Scientific Maybridge
SPB04957EB
25 g
320.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

169814-48-6
235.67
XSAMMAPYCPHHKH-UHFFFAOYSA-N
2735240
CC(=O)C1=C(C)ON=C1C1=CC=C(Cl)C=C1
C12H10ClNO2
MFCD01313507
1-3-4-chlorophenyl-5-methylisoxazol-4-yl ethan-1-one, 1-3-4-chlorophenyl-5-methylisoxazol-4-yl ethanone, 1-3-4-chlorophenyl-5-methyl-1,2-oxazol-4-yl ethanone, 3-4-chlorophenyl-4-acetyl-5-methylisoxazole, ethanone, 1-3-4-chlorophenyl-5-methyl-4-isoxazolyl, maybridge1_008689, 4-acetyl-3-4-chlorophenyl-5-methylisoxazole, 1-3-4-chlorophenyl-5-methyl-4-isoxazolyl-ethanone, 1-3-4-chloro-phenyl-5-methyl-isoxazol-4-yl-ethanone
1-[3-(4-chlorophenyl)-5-methyl-1,2-oxazol-4-yl]ethan-1-one

Specifications

169814-48-6
97%
C12H10ClNO2
1-3-4-chlorophenyl-5-methylisoxazol-4-yl ethan-1-one, 1-3-4-chlorophenyl-5-methylisoxazol-4-yl ethanone, 1-3-4-chlorophenyl-5-methyl-1,2-oxazol-4-yl ethanone, 3-4-chlorophenyl-4-acetyl-5-methylisoxazole, ethanone, 1-3-4-chlorophenyl-5-methyl-4-isoxazolyl, maybridge1_008689, 4-acetyl-3-4-chlorophenyl-5-methylisoxazole, 1-3-4-chlorophenyl-5-methyl-4-isoxazolyl-ethanone, 1-3-4-chloro-phenyl-5-methyl-isoxazol-4-yl-ethanone
CC(=O)C1=C(C)ON=C1C1=CC=C(Cl)C=C1
235.67
235.67
1-[3-(4-Chlorophenyl)-5-methylisoxazol-4-yl]ethan-1-one
97.0
Amber Glass Bottle
MFCD01313507
XSAMMAPYCPHHKH-UHFFFAOYSA-N
1-[3-(4-chlorophenyl)-5-methyl-1,2-oxazol-4-yl]ethan-1-one
2735240
97%
Videos
SDS
Documents

Documents

Product Certifications