missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Benzenetricarboxylic acid chloride, 98%, Thermo Scientific Chemicals

Product Code. 10684642
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4422-95-1
Molecular Formula C9H3Cl3O3
Molecular Weight (g/mol) 265.47
MDL Number MFCD00000679
InChI Key UWCPYKQBIPYOLX-UHFFFAOYSA-N
Synonym trimesoyl chloride, 1,3,5-benzenetricarbonyl trichloride, 1,3,5-benzenetricarboxylic acid chloride, 1,3,5-benzenetricarbonyl chloride, trimesic acid trichloride, 1,3,5-benzenetricarboxylic chloride, benzene-1,3,5-tricarbonyl trichloride, unii-5866peo79w, 1,5-benzenetricarbonyl trichloride, 1,3,5-benzenetricarboxylicacidchloride
PubChem CID 78138
IUPAC Name benzene-1,3,5-tricarbonyl chloride
SMILES ClC(=O)C1=CC(=CC(=C1)C(Cl)=O)C(Cl)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 34.5°C to 36°C
Color Colorless to Yellow
Boiling Point 180°C (16.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97.5 to 102.5% (ex Cl) (Argentometry)
Packaging Glass bottle
Linear Formula C6H3(COCl)3
Refractive Index 1.5945 to 1.5965
Quantity 100 g
Beilstein 09, IV, 3748
Solubility Information Solubility in water: decomposes
Formula Weight 265.47
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1, 3, 5-Benzenetricarboxylic acid chloride
Show More Show Less
compliance-icons
Product Identifier
  • 1,3,5-Benzenetricarboxylic acid chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.