missing translation for 'onlineSavingsMsg'
Learn More
1-(3,5-Dichlorophenyl)-1H-pyrrole-2-carbaldehyde, 97%, Thermo Scientific™ Product Code.: 10572642

1-(3,5-Dichlorophenyl)-1H-pyrrole-2-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10572642
10 g, Amber Glass Bottle
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10572642

Brand: Thermo Scientific Maybridge CD08850EA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175136-79-5
Molecular Formula C11H7Cl2NO
Molecular Weight (g/mol) 240.08
MDL Number MFCD00174281
InChI Key GNBDQGBCNPLAQK-UHFFFAOYSA-N
Synonym 1-3,5-dichlorophenyl-1h-pyrrole-2-carbaldehyde, 1-3,5-dichlorophenyl pyrrole-2-carbaldehyde, 1-3,5-dichlorophenyl-1h-pyrrole-2-carboxaldehyde, acmc-1c802, n-3,5-dichlorophenyl pyrrole-2-carboxaldehyde, 1-3,5-dichlorophenyl pyrrole-2-carboxaldehyde, 1-3,5-dichloro-phenyl-1h-pyrrole-2-carbald ehyde, 1h-pyrrole-2-carboxaldehyde,1-3,5-dichlorophenylShow MoreShow Less
PubChem CID 2777016
SMILES ClC1=CC(=CC(Cl)=C1)N1C=CC=C1C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
IUPAC Name 1-(3,5-dichlorophenyl)pyrrole-2-carbaldehyde
Formula Weight 240.09
Percent Purity 97%
Chemical Name or Material 1-(3, 5-Dichlorophenyl)-1H-pyrrole-2-carbaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-(3,5-Dichlorophenyl)-1H-pyrrole-2-carbaldehyde, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.